siRNA Details
| Gene Symbol: 1700030N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006564.1 | - | 1 | 861 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700030N18Rik| Gene Symbol: 1700030N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006564.1 | - | 1 | 861 | ||