siRNA Details
| Gene Symbol: 6330571C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018247.1 | - | 1 | 1188 | ||
| AV335343.1 | - | 1 | 178 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6330571C24Rik| Gene Symbol: 6330571C24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018247.1 | - | 1 | 1188 | ||
| AV335343.1 | - | 1 | 178 | |||