siRNA Details
| Gene Symbol: Gm20356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040450.1 | 4 | 1404 | |||
| GenBank mRNA | AK139930.1 | - | 4 | 1405 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm20356| Gene Symbol: Gm20356 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040450.1 | 4 | 1404 | |||
| GenBank mRNA | AK139930.1 | - | 4 | 1405 | ||