siRNA Details
| Gene Symbol: 1700063D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040392.1 | 3 | 387 | |||
| GenBank mRNA | AK018871.1 | - | 3 | 384 | ||
| BC048584.1 | - | 3 | 354 | |||
| CJ056044.1 | - | 2 | 345 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU938569.1 | - | 3 | 357 | Chr. 9 - 41206186 - 41217571 [+] on Build GRCm38 | |

1700063D05Rik
Genomic Map