siRNA Details
| Gene Symbol: C030013D06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021073.1 | - | 1 | 474 | ||
| AK087405.1 | - | 2 | 1058 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030013D06Rik| Gene Symbol: C030013D06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021073.1 | - | 1 | 474 | ||
| AK087405.1 | - | 2 | 1058 | |||