siRNA Details
| Gene Symbol: D16Ertd519e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040474.1 | 3 | 527 | |||
| GenBank mRNA | AK139678.1 | - | 3 | 528 | ||
| CO811502.1 | - | 3 | 536 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY725566.1 | - | 3 | 528 | Chr. 16 - 70616425 - 70624822 [+] on Build GRCm38 | |

D16Ertd519e
Genomic Map