siRNA Details
| Gene Symbol: A230103J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110581.1 | 3 | 1475 | |||
| NR_110582.1 | 4 | 1123 | ||||
| GenBank mRNA | AK039159.1 | - | 4 | 1124 | ||
| AK047741.1 | - | 3 | 1444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230103J11Rik| Gene Symbol: A230103J11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110581.1 | 3 | 1475 | |||
| NR_110582.1 | 4 | 1123 | ||||
| GenBank mRNA | AK039159.1 | - | 4 | 1124 | ||
| AK047741.1 | - | 3 | 1444 | |||