siRNA Details
| Gene Symbol: 6030492E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020086.1 | - | 1 | 703 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV073193.1 | - | 247 | Chr. - - [-] on Build | ||

6030492E11Rik
Genomic Map