siRNA Details
| Gene Symbol: D10Bwg1070e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017596.1 | - | 1 | 28 | ||
| AK156477.1 | - | 1 | 941 | |||
| AK158707.1 | - | 1 | 877 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D10Bwg1070e| Gene Symbol: D10Bwg1070e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017596.1 | - | 1 | 28 | ||
| AK156477.1 | - | 1 | 941 | |||
| AK158707.1 | - | 1 | 877 | |||