siRNA Details
| Gene Symbol: C030009O12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021058.1 | - | 1 | 184 | ||
| AK045829.1 | - | 1 | 608 | |||
| AK140124.1 | - | 1 | 658 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB284330.1 | - | 1 | 81 | Chr. 5 - 72146987 - 72147501 [+] on Build GRCm38 | |

C030009O12Rik
Genomic Map