siRNA Details
| Gene Symbol: Gm3916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA465826.1 | - | 2 | 91 | ||
| AK086006.1 | - | 2 | 151 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB662335.1 | - | 2 | 148 | Chr. 19 - 27260732 - 27262321 [-] on Build GRCm38 | |

Gm3916
Genomic Map