siRNA Details
| Gene Symbol: 9530014B07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA671975.1 | - | 2 | 88 | ||
| AK020556.1 | - | 2 | 83 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722006.1 | - | 2 | 83 | Chr. 13 - 53635982 - 53639193 [-] on Build GRCm38 | |

9530014B07Rik
Genomic Map