siRNA Details
| Gene Symbol: Gm30498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110507.1 | 6 | 573 | |||
| GenBank mRNA | BC049705.1 | - | 6 | 573 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU939480.1 | - | 576 | Chr. - - [-] on Build | ||

Gm30498
Genomic Map