siRNA Details
| Gene Symbol: Plcb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020944.1 | - | 2 | 522 | ||
| AK080865.1 | - | 2 | 789 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB323656.2 | - | 1 | 90 | Chr. 2 - 135742100 - 135742751 [+] on Build GRCm38 | |

Plcb4
Genomic Map