siRNA Details
Gene Symbol: LOC105376114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135839.1 | 2 | 740 | |||
NR_135840.1 | 2 | 385 | ||||
GenBank mRNA | AK300656.1 | - | 2 | 377 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105376114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135839.1 | 2 | 740 | |||
NR_135840.1 | 2 | 385 | ||||
GenBank mRNA | AK300656.1 | - | 2 | 377 |