siRNA Details
| Gene Symbol: C5orf17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_131245.1 | 2 | 478 | |||
| GenBank mRNA | AK092155.1 | - | 2 | 478 | ||
| BQ950913.1 | - | 2 | 438 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000512559.5 | - | 2 | 478 | Chr. 5 - 23951348 - 24178263 [+] on Build GRCh38 | |

C5orf17
Genomic Map