siRNA Details
| Gene Symbol: LINC01558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB016901.1 | - | 3 | 1036 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000366822.2 | - | 3 | 1011 | Chr. 6 - 167786820 - 167796859 [-] on Build GRCh38 | |

LINC01558
Genomic Map