siRNA Details
| Gene Symbol: LOC101927342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125939.1 | 1, 2 | 73 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927342| Gene Symbol: LOC101927342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125939.1 | 1, 2 | 73 | |||