This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LINC01529 SIRNA Assays ›
Gene Symbol:
LINC01529
Gene Name:
long intergenic non-protein coding RNA 1529
Gene Aliases:
-
Chromosome Location:
Chr.19: 35788876 - 35797909 on Build GRCh38
UniGene ID:
Hs.651888
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n504920

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 392.00
Your Price
Online offer: 380.65
380.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINC01529
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_104176.1 6 1711
GenBank mRNA AK055260.1 - 6 1687

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0