siRNA Details
| Gene Symbol: UBXN10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110078.1 | 2 | 150 | |||
| GenBank mRNA | BX106299.1 | - | 2 | 137 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UBXN10-AS1| Gene Symbol: UBXN10-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110078.1 | 2 | 150 | |||
| GenBank mRNA | BX106299.1 | - | 2 | 137 | ||