siRNA Details
| Gene Symbol: LOC101926955 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110003.1 | 4 | 2002 | |||
| GenBank mRNA | BC052952.1 | - | 4 | 2002 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000449316.1 | - | 4 | 2002 | Chr. 20 - 25955812 - 25969288 [+] on Build GRCh38 | |

LOC101926955
Genomic Map