siRNA Details
| Gene Symbol: LOC102724094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120519.1 | 1 | 29 | |||
| NR_120520.1 | 1 | 29 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000448513.1 | - | 1 | 22 | Chr. 7 - 101014325 - 101017601 [-] on Build GRCh38 | |

LOC102724094
Genomic Map