siRNA Details
| Gene Symbol: DNAJC9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134459.1 | 2 | 547 | |||
| NR_134460.1 | 3 | 579 | ||||
| GenBank mRNA | BC033983.1 | - | 2 | 502 | ||
| BC047404.1 | - | 1 | 1374 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAJC9-AS1| Gene Symbol: DNAJC9-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134459.1 | 2 | 547 | |||
| NR_134460.1 | 3 | 579 | ||||
| GenBank mRNA | BC033983.1 | - | 2 | 502 | ||
| BC047404.1 | - | 1 | 1374 | |||