siRNA Details
| Gene Symbol: LINC00858 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038220.1 | 5 | 1024 | |||
| GenBank mRNA | AK056561.1 | - | 5 | 1024 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000415469.5 | - | 5 | 1024 | Chr. 10 - 84279980 - 84294659 [+] on Build GRCh38 | |

LINC00858
Genomic Map