siRNA Details
| Gene Symbol: LOC105376365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134490.1 | 2 | 440 | |||
| GenBank mRNA | BM720503.1 | - | 2 | 440 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000413339.1 | - | 2 | 440 | Chr. 10 - 3833950 - 3834728 [+] on Build GRCh38 | |

LOC105376365
Genomic Map