siRNA Details
| Gene Symbol: LOC101926941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120664.1 | 1, 2 | 86 | |||
| GenBank mRNA | AI700977.1 | - | 1, 2 | 81 | ||
| DA537977.1 | - | 1, 2 | 86 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101926941| Gene Symbol: LOC101926941 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120664.1 | 1, 2 | 86 | |||
| GenBank mRNA | AI700977.1 | - | 1, 2 | 81 | ||
| DA537977.1 | - | 1, 2 | 86 | |||