siRNA Details
| Gene Symbol: LINC01307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126402.1 | 4 | 302 | |||
| GenBank mRNA | BG203915.1 | - | 1 | 23 | ||
| W96233.1 | - | 4 | 301 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000444327.1 | - | 1 | 21 | Chr. 1 - 101350049 - 101390303 [+] on Build GRCh38 | |

LINC01307
Genomic Map