siRNA Details
| Gene Symbol: LINC01412 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110570.1 | 2 | 178 | |||
| GenBank mRNA | HY038051.1 | - | 179 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000413525.1 | - | 1 | 117 | Chr. 2 - 144566435 - 144579434 [+] on Build GRCh38 | |

LINC01412
Genomic Map