siRNA Details
| Gene Symbol: GACAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126369.1 | 2, 3 | 287 | |||
| NR_126370.1 | 2, 3 | 287 | ||||
| GenBank mRNA | DB077273.1 | - | 2, 3 | 254 | ||
| DB096621.1 | - | 2, 3 | 287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GACAT1| Gene Symbol: GACAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126369.1 | 2, 3 | 287 | |||
| NR_126370.1 | 2, 3 | 287 | ||||
| GenBank mRNA | DB077273.1 | - | 2, 3 | 254 | ||
| DB096621.1 | - | 2, 3 | 287 | |||