siRNA Details
| Gene Symbol: LOC727993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134520.1 | 4 | 1494 | |||
| GenBank mRNA | AK090681.1 | - | 4 | 1461 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000446562.1 | - | 4 | 1458 | Chr. 20 - 1947246 - 2007517 [+] on Build GRCh38 | |

LOC727993
Genomic Map