siRNA Details
| Gene Symbol: LINC01440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110629.1 | 1 | 73 | |||
| GenBank mRNA | BI464301.1 | - | 1 | 77 | ||
| BX107747.1 | - | 1 | 51 | |||
| DB449830.1 | - | 1 | 73 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01440| Gene Symbol: LINC01440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110629.1 | 1 | 73 | |||
| GenBank mRNA | BI464301.1 | - | 1 | 77 | ||
| BX107747.1 | - | 1 | 51 | |||
| DB449830.1 | - | 1 | 73 | |||