siRNA Details
| Gene Symbol: NEURL1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120676.1 | 3 | 189 | |||
| GenBank mRNA | BX095052.1 | - | 3 | 163 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000447820.1 | - | 2 | 132 | Chr. 10 - 103511779 - 103515402 [-] on Build GRCh38 | |

NEURL1-AS1
Genomic Map