This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BHLHE40-AS1 SIRNA Assays ›
Gene Symbol:
BHLHE40-AS1
Gene Name:
BHLHE40 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 4896809 - 4979961 on Build GRCh38
UniGene ID:
Hs.434505
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID n508336

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BHLHE40-AS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037903.3 3 891
NR_125914.1 3 850
NR_125915.1 4 795
NR_125916.1 3 655
GenBank mRNA AK056892.1 - 4 795
AK311159.1 - 3 850
AK311646.1 - 3 867
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000620618.4 - 795 Chr. - - [-] on Build
GENCODE ENST00000615017.4 - 850 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0