siRNA Details
| Gene Symbol: LINC01282 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110385.1 | 6 | 852 | |||
| GenBank mRNA | BC043536.1 | - | 6 | 852 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000431646.1 | - | 6 | 852 | Chr. X - 39367285 - 39391774 [-] on Build GRCh38 | |

LINC01282
Genomic Map