siRNA Details
| Gene Symbol: ZNF503-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024421.1 | 2 | 1356 | |||
| NR_024422.1 | 2 | 924 | ||||
| NR_110299.1 | 2 | 920 | ||||
| NR_110300.1 | 2 | 692 | ||||
| NR_110301.1 | 2 | 809 | ||||
| NR_110302.1 | 2 | 742 | ||||
| GenBank mRNA | BC029963.1 | - | 2 | 692 | ||

ZNF503-AS2
Genomic Map