siRNA Details
| Gene Symbol: LOC101929653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110140.1 | 7 | 2006 | |||
| GenBank mRNA | AK128059.1 | - | 7 | 2006 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929653| Gene Symbol: LOC101929653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110140.1 | 7 | 2006 | |||
| GenBank mRNA | AK128059.1 | - | 7 | 2006 | ||