siRNA Details
| Gene Symbol: LOC101929653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110140.1 | 5 | 653 | |||
| GenBank mRNA | AK128059.1 | - | 5 | 653 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000414466.5 | - | 5 | 653 | Chr. 11 - 131533857 - 131540867 [-] on Build GRCh38 | |

LOC101929653
Genomic Map