siRNA Details
| Gene Symbol: LINC00971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033860.1 | 19 | 4450 | |||
| GenBank mRNA | BC068246.1 | - | 19 | 4453 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000484892.5 | - | 19 | 4450 | Chr. 3 - 84638406 - 84869575 [-] on Build GRCh38 | |

LINC00971
Genomic Map