siRNA Details
| Gene Symbol: LOC101928445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110560.1 | 9 | 1459 | |||
| GenBank mRNA | AK128666.1 | - | 9 | 1459 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500262.1 | - | 9 | 1459 | Chr. 5 - 179595904 - 179603741 [-] on Build GRCh38 | |

LOC101928445
Genomic Map