siRNA Details
| Gene Symbol: LINC01252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033890.1 | 8 | 3431 | |||
| GenBank mRNA | BC041827.1 | - | 8 | 3434 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000499291.6 | - | 8 | 3431 | Chr. 12 - 11548030 - 11564403 [+] on Build GRCh38 | |

LINC01252
Genomic Map