siRNA Details
| Gene Symbol: LOC102467147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104664.1 | 4 | 465 | |||
| GenBank mRNA | BX484633.1 | - | 4 | 466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102467147| Gene Symbol: LOC102467147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104664.1 | 4 | 465 | |||
| GenBank mRNA | BX484633.1 | - | 4 | 466 | ||