siRNA Details
| Gene Symbol: LOC100506272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110436.1 | 3 | 406 | |||
| GenBank mRNA | BX108713.1 | - | 3 | 407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100506272| Gene Symbol: LOC100506272 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110436.1 | 3 | 406 | |||
| GenBank mRNA | BX108713.1 | - | 3 | 407 | ||