siRNA Details
| Gene Symbol: PGGHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011520383.2 | XP_011518685.1 | 3 | 601 | ||
| XM_011520384.2 | XP_011518686.1 | 2 | 712 | |||
| XM_011520385.2 | XP_011518687.1 | 2 | 715 | |||
| XM_011520386.2 | XP_011518688.1 | 2 | 715 | |||
| XM_011520387.2 | XP_011518689.1 | 2 | 715 | |||
| XM_017018355.1 | XP_016873844.1 | 1 | 65 | |||

PGGHG
Genomic Map