siRNA Details
| Gene Symbol: FLJ41278 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033988.1 | 3 | 2889 | |||
| GenBank mRNA | AK123272.1 | - | 3 | 2894 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000535058.1 | - | 3 | 3269 | Chr. 12 - 64883394 - 64977522 [+] on Build GRCh38 | |

FLJ41278
Genomic Map