siRNA Details
| Gene Symbol: LINC00517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135286.1 | 2 | 1440 | |||
| GenBank mRNA | AK125955.1 | - | 2 | 1441 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000554257.1 | - | 2 | 1440 | Chr. 14 - 37896060 - 37902124 [-] on Build GRCh38 | |

LINC00517
Genomic Map