siRNA Details
| Gene Symbol: LOC374890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135233.1 | 2 | 186 | |||
| GenBank mRNA | BC030765.2 | - | 2 | 186 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000598042.1 | - | 2 | 186 | Chr. 19 - 22615557 - 22623971 [-] on Build GRCh38 | |

LOC374890
Genomic Map