siRNA Details
| Gene Symbol: LOC105370333 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132422.1 | 1 | 282 | |||
| GenBank mRNA | AV692313.1 | - | 1 | 282 | ||
| BG033002.1 | - | 1 | 268 | |||
| CD248433.1 | - | 1 | 219 | |||
| CF454479.1 | - | 1 | 229 | |||
| CX784348.1 | - | 1 | 187 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000612598.1 | - | 187 | Chr. - - [-] on Build | ||

LOC105370333
Genomic Map