siRNA Details
| Gene Symbol: BANCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047671.2 | 3 | 392 | |||
| GenBank mRNA | N98863.1 | - | 3 | 301 | ||
| W58600.1 | - | 3 | 234 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000624238.1 | - | 194 | Chr. - - [-] on Build | ||

BANCR
Genomic Map