siRNA Details
| Gene Symbol: GPR1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104359.1 | 6 | 1675 | |||
| GenBank mRNA | AK002005.1 | - | 1 | 147 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000623040.1 | - | 1675 | Chr. - - [-] on Build | ||

GPR1-AS
Genomic Map