siRNA Details
| Gene Symbol: LOC101929608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109878.1 | 4 | 642 | |||
| GenBank mRNA | AK310670.1 | - | 4 | 642 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000624692.1 | - | 642 | Chr. - - [-] on Build | ||

LOC101929608
Genomic Map